4-Chloro-3-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Chloro-3-methoxyphenyl)methanol

26519-1G 26519-25G 26519-5G
76929-210EA 70.02 USD
76929-210 76929-212 76929-214
4-Chloro-3-methoxybenzyl alcohol ≥97%
4-Chloro-3-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
MW: 172.609 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15527076
CAS Number: 13726-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR