4-Chloro-3-methoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

26438-1G 26438-25G 26438-5G
76929-196EA 111.4 USD
76929-196 76929-198 76929-200
4-Chloro-3-methoxybenzonitrile ≥97%
4-Chloro-3-methoxybenzonitrile
Formula: C₈H₆ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD09753729
CAS Number: 189628-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR