4-Hydroxymethyl-3-methoxyphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

26377-1G 26377-25G 26377-500MG 26377-5G
76929-182EA 175.05 USD
76929-182 76929-184 76929-186 76929-188
4-Hydroxymethyl-3-methoxyphenylboronic acid pinacol ester ≥95%
4-Hydroxymethyl-3-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₄
MW: 264.125 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18732626
CAS Number: 1404094-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR