Methyl 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
Benzoic acid, 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, methyl ester, 4-Fluoro-3-(methoxycarbonyl)phenylboronic acid pinacol ester, 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester
26321-25G
26321-5G
76929-176EA
357.53
USD
76929-176
76929-178
Methyl 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Methyl 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Precaución: for Research Use Only
Formula:
C₁₄H₁₈BFO₄ MW: 280.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16996316 CAS Number: 872459-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...