4-Methoxy-2,6-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

26297-1G 26297-25G 26297-5G
76929-166EA 105.03 USD
76929-166 76929-168 76929-170
4-Methoxy-2,6-dimethylphenol ≥97%
4-Methoxy-2,6-dimethylphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD14537195
CAS Number: 2431-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR