4-Chloro-3,5-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

26238-25G 26238-5G
76929-128EA 488.02 USD
76929-128 76929-130
4-Chloro-3,5-difluorobenzaldehyde ≥97%
4-Chloro-3,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD11553555
CAS Number: 1160573-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR