4'-Bromo-4-(trifluoromethoxy)biphenyl ≥97%

Supplier: AOBChem USA

Warning

26206-1G 26206-25G 26206-500MG 26206-5G
76929-090EA 206.88 USD
76929-090 76929-092 76929-094 76929-096
4'-Bromo-4-(trifluoromethoxy)biphenyl ≥97%
4'-Bromo-4-(trifluoromethoxy)biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₈BrF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD04972698
CAS Number: 134150-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR