4-Ethoxy-3-fluorophenol ≥95% research grade

Supplier: AOBChem USA

Warning

26121-1G 26121-25G 26121-5G
76929-070EA 84.88 USD
76929-070 76929-072 76929-074
4-Ethoxy-3-fluorophenol ≥95% research grade
4-Ethoxy-3-fluorophenol
Formula: C₈H₉FO₂
MW: 156.154 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21984328
CAS Number: 98121-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR