5-Bromo-2-(trifluoromethoxy)phenol ≥97%

Supplier: AOBChem USA

Warning

26060-1G 26060-25G 26060-5G
76929-022EA 66.84 USD
76929-022 76929-024 76929-026
5-Bromo-2-(trifluoromethoxy)phenol ≥97%
5-Bromo-2-(trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O₂
MW: 257.005 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21333031
CAS Number: 1048963-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR