4-Bromo-5-fluoro-2-nitroaniline ≥97%

Supplier: AOBChem USA

Warning

26010-25G 26010-5G
76929-010EA 240.83 USD
76929-010 76929-012
4-Bromo-5-fluoro-2-nitroaniline ≥97%
4-Bromo-5-fluoro-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrFN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11101433
CAS Number: 153505-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR