5-Bromo-7-chloro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

25952-1G 25952-25G 25952-5G
76928-996EA 62.6 USD
76928-996 76928-998 76929-000
5-Bromo-7-chloro-1H-indazole ≥97%
5-Bromo-7-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.4771 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044588
CAS Number: 635712-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR