5-Bromo-2-fluoro-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

25936-1G 25936-25G 25936-5G
76928-982EA 70.02 USD
76928-982 76928-984 76928-986
5-Bromo-2-fluoro-3-methylbenzyl alcohol ≥97%
5-Bromo-2-fluoro-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24848695
CAS Number: 1197944-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR