4-Methoxy-3-methylbenzyl alcohol ≥95% research grade

Supplier: AOBChem USA

Warning

25865-1G 25865-25G 25865-5G
76928-968EA 62.6 USD
76928-968 76928-970 76928-972
4-Methoxy-3-methylbenzyl alcohol ≥95% research grade
4-Methoxy-3-methylbenzyl alcohol
Formula: C₉H₁₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00060357
CAS Number: 114787-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR