5-Bromo-3-chloro-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

25861-1G 25861-25G 25861-500MG 25861-5G
76928-956EA 231.28 USD
76928-956 76928-958 76928-960 76928-962
5-Bromo-3-chloro-2-fluorobenzoic acid ≥97%
5-Bromo-3-chloro-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24107274
CAS Number: 1449008-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR