5-Chloro-2-hydrazinopyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-2-hydrazinylpyridine

25841-25G 25841-5G
76928-940EA 183.54 USD
76928-940 76928-942
5-Chloro-2-hydrazinopyridine ≥97%
5-Chloro-2-hydrazinopyridine
Formula: C₅H₆ClN₃
MW: 143.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07786313
CAS Number: 27032-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR