(2-Methyl-4-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Methoxy-2-methylbenzyl alcohol

25833-1G 25833-25G 25833-500MG 25833-5G
76928-928EA 153.84 USD
76928-928 76928-930 76928-932 76928-934
(2-Methyl-4-methoxyphenyl)methanol ≥97%
(2-Methyl-4-methoxyphenyl)methanol
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04038833
CAS Number: 52289-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR