4-Methoxy-3-(1-methylethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

25827-1G 25827-25G 25827-500MG 25827-5G
76928-918EA 290.69 USD
76928-918 76928-920 76928-922 76928-924
4-Methoxy-3-(1-methylethyl)phenylboronic acid ≥97%
4-Methoxy-3-(1-methylethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MW: 194.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414591
CAS Number: 290348-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR