4-Hydroxy-3-isopropylphenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Hydroxy-3-isopropylphenylboronic acid pinacol ester, 2-Propan-2-yl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

25822-1G 25822-25G 25822-500MG 25822-5G
76928-904EA 245.07 USD
76928-904 76928-906 76928-908 76928-910
4-Hydroxy-3-isopropylphenylboronic acid, pinacol ester ≥97%
4-Hydroxy-3-isopropylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₃
MW: 262.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16994212
CAS Number: 1188335-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR