5-Bromo-2-iodo-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

25714-1G 25714-25G 25714-500MG 25714-5G
76928-878EA 126.25 USD
76928-878 76928-880 76928-882 76928-884
5-Bromo-2-iodo-4-methylpyridine ≥97%
5-Bromo-2-iodo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrIN
Storage Temperature: Refrigerator
MDL Number: MFCD09801049
CAS Number: 941294-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR