5-Bromo-3-hydroxy-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-methoxypyridin-3-ol, 5-Bromo-3-hydroxypyridin-2-yl methyl ether

25674-1G 25674-25G 25674-500MG 25674-5G
76928-862EA 228.1 USD
76928-862 76928-864 76928-866 76928-868
5-Bromo-3-hydroxy-2-methoxypyridine ≥97%
5-Bromo-3-hydroxy-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂
MW: 204.021 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18256269
CAS Number: 1211589-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR