5-Chloro-2,4-difluorobenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

25594-1G 25594-25G 25594-5G
76928-836EA 96.55 USD
76928-836 76928-838 76928-840
5-Chloro-2,4-difluorobenzyl alcohol ≥95%
5-Chloro-2,4-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅ClF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD22200072
CAS Number: 915409-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR