5-Chloro-2,4-difluorobromobenzene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-5-chloro-2,4-difluorobenzene, 5-Bromo-1-chloro-2,4-difluorobenzene

25583-25G 25583-5G
76928-830EA 283.27 USD
76928-830 76928-832
5-Chloro-2,4-difluorobromobenzene ≥97% research grade
5-Chloro-2,4-difluorobromobenzene
Formula: C₆H₂BrClF₂
MW: 227.434 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112557
CAS Number: 914636-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR