4-Methyl-1H-indazole ≥95%
Supplier: AOBChem USA
|
Warning
|
25418-25G
25418-5G
76928-782EA
346.92
USD
76928-782
76928-784
4-Methyl-1H-indazole ≥95%
4-Methyl-1H-indazole
Precaución: for Research Use Only
Formula:
C₈H₈N₂ MW: 132.1600 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06200231 CAS Number: 3176-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...