1-Chloro-2,5-difluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2,5-difluoroiodobenzene

25327-25G 25327-5G
76928-744EA 403.15 USD
76928-744 76928-746
1-Chloro-2,5-difluoro-4-iodobenzene ≥97%
1-Chloro-2,5-difluoro-4-iodobenzene
Formula: C₆H₂ClF₂I
MW: 274.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18379746
CAS Number: 1097871-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR