4-Bromo-3,5-dichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

25286-1G 25286-25G 25286-500MG 25286-5G
76928-730EA 213.25 USD
76928-730 76928-732 76928-734 76928-736
4-Bromo-3,5-dichlorobenzaldehyde ≥97%
4-Bromo-3,5-dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD19686182
CAS Number: 120077-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR