4-Bromo-2-isopropylphenol ≥97%

Supplier: AOBChem USA

Warning

25258-25G 25258-5G
76928-708EA 630.18 USD
76928-708 76928-710
4-Bromo-2-isopropylphenol ≥97%
4-Bromo-2-isopropylphenol
Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD00040203
CAS Number: 26307-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR