4-Bromo-5-chloro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

25217-1G 25217-25G 25217-500MG 25217-5G
76928-692EA 379.81 USD
76928-692 76928-694 76928-696 76928-698
4-Bromo-5-chloro-2-methylbenzoic acid ≥97%
4-Bromo-5-chloro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18917161
CAS Number: 1349716-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR