4-Methyl-3-phenylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

25177-1G 25177-25G 25177-500MG 25177-5G
76928-676EA 168.69 USD
76928-676 76928-678 76928-680 76928-682
4-Methyl-3-phenylpyridine ≥97% research grade
4-Methyl-3-phenylpyridine
Formula: C₁₂H₁₁N
Storage Temperature: Refrigerator
MDL Number: MFCD04114238
CAS Number: 19352-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR