5-Cyclopropylpyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

25126-1G 25126-25G 25126-500MG 25126-5G
76928-660EA 335.25 USD
76928-660 76928-662 76928-664 76928-666
5-Cyclopropylpyridine-3-boronic acid pinacol ester ≥97%
5-Cyclopropylpyridine-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₂
MW: 245.125 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13182162
CAS Number: 1220696-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR