5-Hydroxy-2-methoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
25106-1G
25106-25G
25106-500MG
25106-5G
76928-644EA
320.4
USD
76928-644
76928-646
76928-648
76928-650
5-Hydroxy-2-methoxybenzoic acid ≥97%
5-Hydroxy-2-methoxybenzoic acid
Formula:
C₈H₈O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD10698756 CAS Number: 61227-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...