4-Fluoro-2-iodoanisole ≥95%

Supplier: AOBChem USA

Warning

25025-1G 25025-25G 25025-500MG 25025-5G
76928-628EA 228.1 USD
76928-628 76928-630 76928-632 76928-634
4-Fluoro-2-iodoanisole ≥95%
4-Fluoro-2-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD00464332
CAS Number: 3824-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR