5-Bromo-6-chloro-1H-indazole ≥97% research grade

Supplier: AOBChem USA

Warning

25012-1G 25012-25G 25012-5G
76928-614EA 96.55 USD
76928-614 76928-616 76928-618
5-Bromo-6-chloro-1H-indazole ≥97% research grade
5-Bromo-6-chloro-1H-indazole
Formula: C₇H₄BrClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD18381158
CAS Number: 1260382-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR