5-Bromo-2-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Bromo-2-methylphenyl)methanol

24936-25G 24936-5G
76928-602EA 466.8 USD
76928-602 76928-604
5-Bromo-2-methylbenzyl alcohol ≥97%
5-Bromo-2-methylbenzyl alcohol
Formula: C₈H₉BrO
MW: 201.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12031921
CAS Number: 258886-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR