5-Cyclopropyl-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

24888-25G 24888-5G
76928-566EA 418 USD
76928-566 76928-568
5-Cyclopropyl-2-fluoropyridine ≥97%
5-Cyclopropyl-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₈H₈FN
Storage Temperature: Refrigerator
MDL Number: MFCD18384289
CAS Number: 1034467-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR