(4-Bromo-2-isopropylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-isopropylphenylboronic acid

24881-1G 24881-25G 24881-500MG 24881-5G
76928-550EA 350.1 USD
76928-550 76928-552 76928-554 76928-556
(4-Bromo-2-isopropylphenyl)boronic acid ≥97% research grade
(4-Bromo-2-isopropylphenyl)boronic acid
Formula: C₉H₁₂BBrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD22628234
CAS Number: 1451390-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR