4-Fluoro-3-(methylthio)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

24750-1G 24750-25G 24750-500MG 24750-5G
76928-508EA 409.51 USD
76928-508 76928-510 76928-512 76928-514
4-Fluoro-3-(methylthio)benzyl alcohol ≥97%
4-Fluoro-3-(methylthio)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FOS
MW: 172.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24848712
CAS Number: 1449008-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR