5-Bromo-3-ethoxy-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

24730-1G 24730-25G 24730-500MG 24730-5G
76928-494EA 144.29 USD
76928-494 76928-496 76928-498 76928-500
5-Bromo-3-ethoxy-2-fluoropyridine ≥97%
5-Bromo-3-ethoxy-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₇H₇BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD22628210
CAS Number: 1414870-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR