5-Bromo-1-methylpyridin-2(1H)-one ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-1-methyl-2(1H)-pyridinone

24706-25G 24706-5G
76928-482EA 254.62 USD
76928-482 76928-484
5-Bromo-1-methylpyridin-2(1H)-one ≥97%
5-Bromo-1-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972188
CAS Number: 81971-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR