4-Cyclopropyl-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

24647-1G 24647-25G 24647-500MG 24647-5G
76928-470EA 307.67 USD
76928-470 76928-472 76928-474 76928-476
4-Cyclopropyl-2-methylphenylboronic acid ≥97%
4-Cyclopropyl-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD21099610
CAS Number: 2121514-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR