5-Bromo-4-methyl-1-(2-methylpropoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

24452-1G 24452-25G 24452-500MG 24452-5G
76928-390EA 260.99 USD
76928-390 76928-392 76928-394 76928-396
5-Bromo-4-methyl-1-(2-methylpropoxy)pyridine ≥97%
5-Bromo-4-methyl-1-(2-methylpropoxy)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrNO
Storage Temperature: Ambient
MDL Number: MFCD22628194
CAS Number: 1289131-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR