5-Bromo-2-iodoisopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

24443-25G 24443-5G
76928-382EA 524.09 USD
76928-382 76928-384
5-Bromo-2-iodoisopropylbenzene ≥95%
5-Bromo-2-iodoisopropylbenzene
Formula: C₉H₁₀BrI
MW: 324.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845959
CAS Number: 1147014-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR