Methyl 5-cyano-2-fluorobenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Cyano-2-fluorobenzoic acid methyl ester

24301-25G 24301-5G
76928-330EA 215.37 USD
76928-330 76928-332
Methyl 5-cyano-2-fluorobenzoate ≥97% research grade
Methyl 5-cyano-2-fluorobenzoate
Formula: C₉H₆FNO₂
MW: 179.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042790
CAS Number: 337362-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR