5-Bromo-2-phenoxypyridine ≥97%

Supplier: AOBChem USA

Warning

23980-25G 23980-5G
76928-274EA 275.84 USD
76928-274 76928-276
5-Bromo-2-phenoxypyridine ≥97%
5-Bromo-2-phenoxypyridine

Precaución: for Research Use Only

Formula: C₁₁H₈BrNO
MW: 250.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12761236
CAS Number: 59717-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR