4-Bromo-2-isopropyl-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

23887-1G 23887-25G 23887-500MG 23887-5G
76928-228EA 374.5 USD
76928-228 76928-230 76928-232 76928-234
4-Bromo-2-isopropyl-N-methylbenzamide ≥97%
4-Bromo-2-isopropyl-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO
MW: 256.139 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32631790
CAS Number: 2364584-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR