5-Chloro-2-fluoro-3-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

23839-1G 23839-25G 23839-500MG 23839-5G
76928-204EA 171.87 USD
76928-204 76928-206 76928-208 76928-210
5-Chloro-2-fluoro-3-(trifluoromethyl)benzoic acid ≥95%
5-Chloro-2-fluoro-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃ClF₄O₂
MW: 242.555 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28752591
CAS Number: 1807119-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR