4-Isopropoxy-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

23757-1G 23757-25G 23757-5G
76928-178EA 89.12 USD
76928-178 76928-180 76928-182
4-Isopropoxy-3-methylbenzaldehyde ≥97%
4-Isopropoxy-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12547898
CAS Number: 199743-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR