4-Fluoro-3-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

23603-1G 23603-25G 23603-500MG 23603-5G
76928-132EA 224.92 USD
76928-132 76928-134 76928-136 76928-138
4-Fluoro-3-(methylthio)benzaldehyde ≥95%
4-Fluoro-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FOS
MW: 170.204 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543662
CAS Number: 1289042-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR