4-Fluoro-3-(Methylsulfanyl)Benzoic Acid ≥97%

Supplier: AOBChem USA

Warning

23601-1G 23601-25G 23601-500MG 23601-5G
76928-118EA 409.51 USD
76928-118 76928-120 76928-122 76928-124
4-Fluoro-3-(Methylsulfanyl)Benzoic Acid ≥97%
4-Fluoro-3-(Methylsulfanyl)Benzoic Acid

Precaución: for Research Use Only

Formula: C₈H₇FO₂S
MW: 186.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20657082
CAS Number: 169310-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR