5-Bromo-N,N,4-trimethyl-2-pyridinamine ≥97%

Supplier: AOBChem USA

Warning

23499-1G 23499-25G 23499-500MG 23499-5G
76928-068EA 140.04 USD
76928-068 76928-070 76928-072 76928-074
5-Bromo-N,N,4-trimethyl-2-pyridinamine ≥97%
5-Bromo-N,N,4-trimethyl-2-pyridinamine
Formula: C₈H₁₁BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD08272097
CAS Number: 764651-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR