4-Iodo-2-isopropoxypyridine ≥95%

Supplier: AOBChem USA

Warning

23428-1G 23428-25G 23428-5G
76928-042EA 112.46 USD
76928-042 76928-044 76928-046
4-Iodo-2-isopropoxypyridine ≥95%
4-Iodo-2-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₁₀INO
Storage Temperature: Ambient
MDL Number: MFCD23162752
CAS Number: 1353777-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR